Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027425.3 | NP_081701.2 | 9, 10 | 936 | ||
GenBank mRNA | AK011847.1 | - | 9, 10 | 906 | ||
AK083205.1 | - | 9, 10 | 921 | |||
AK173195.1 | - | 9, 10 | 897 | |||
BC017645.1 | - | 9, 10 | 898 | |||
BC038265.1 | - | 8, 9 | 807 | |||
BC137736.1 | - | 9, 10 | 885 | 8860755 | ||
BC137737.1 | - | 9, 10 | 885 | 8860756 |