Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000277.1 | NP_000268.1 | 6 | 1023 | ||
XM_017019370.1 | XP_016874859.1 | 6 | 666 | |||
GenBank mRNA | AK298419.1 | - | 6 | 647 | ||
AK313383.1 | - | 7 | 648 | |||
K03020.1 | - | 6 | 773 | |||
L47726.1 | - | 6 | 1023 | |||
U49897.1 | - | 6 | 1023 |