Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027444.3 | NP_081720.2 | 5 | 611 | ||
GenBank mRNA | AF276950.1 | - | 5 | 496 | ||
AF276951.1 | - | 5 | 583 | |||
AF276952.1 | - | 2 | 339 | |||
AF454943.1 | - | 5 | 604 | |||
AF454944.1 | - | 3 | 476 | |||
AK017487.1 | - | 5 | 604 | |||
AK029532.1 | - | 3 | 478 | |||
AK029747.1 | - | 6 | 725 | |||
AK049516.1 | - | 5 | 597 | |||
AK050488.1 | - | 5 | 550 | |||
AK157813.1 | - | 5 | 590 | |||
AK167724.1 | - | 5 | 742 | |||
BC019456.1 | - | 5 | 686 | |||
BC024449.1 | - | 3 | 459 | |||
BC038874.1 | - | 1 | 150 | |||
BC057869.1 | - | 5 | 579 |