Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141971.1 | NP_001135443.1 | 4 | 820 | ||
NM_027478.3 | NP_081754.2 | 4 | 880 | |||
GenBank mRNA | AK017729.1 | - | 4 | 874 | ||
BC080755.1 | - | 3 | 2244 | |||
BQ922165.1 | - | 4 | 784 |