Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290701.1 | NP_001277630.1 | 7 | 1026 | ||
NM_027310.4 | NP_081586.1 | 6 | 714 | |||
GenBank mRNA | AK009177.1 | - | 6 | 702 | ||
AK012867.1 | - | 7 | 998 | |||
BC108404.1 | - | 7 | 997 | 30135798 |