Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mospd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290514.1 | NP_001277443.1 | 6 | 1052 | ||
NM_027409.5 | NP_081685.1 | 6 | 1055 | |||
GenBank mRNA | AK007533.1 | - | 6 | 989 | ||
AK139795.1 | - | 6 | 996 | |||
AK203602.1 | - | 2 | 303 | |||
BC018329.1 | - | 6 | 953 | 3962906 | ||
BC085100.1 | - | 6 | 960 | 6391017 | ||
BC100442.1 | - | 6 | 940 | 30501889 | ||
DV651027.1 | - | 3 | 422 |