Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027421.2 | NP_081697.2 | 24 | 3723 | ||
GenBank mRNA | AK013101.1 | - | 5 | 705 | ||
AK129323.1 | - | 24 | 3705 | |||
BC050081.1 | - | 24 | 3499 | 30023969 | ||
BC059056.1 | - | 23 | 3575 | 6821329 | ||
BC118007.1 | - | 24 | 3438 | 40086155 |