Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027432.3 | NP_081708.1 | 10 | 1522 | ||
GenBank mRNA | AK032897.1 | - | 8 | 1061 | ||
AK048488.1 | - | 10 | 1545 | |||
AK050391.1 | - | 10 | 1522 | |||
AK078856.1 | - | 11 | 1481 | |||
AK131608.1 | - | 9 | 1433 | |||
AK161812.1 | - | 9 | 1980 | |||
AK168006.1 | - | 10 | 1523 | |||
AK168752.1 | - | 10 | 1544 | |||
AK169128.1 | - | 10 | 1523 | |||
BC005755.1 | - | 10 | 1477 | 2654151 |