Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027444.3 | NP_081720.2 | 11 | 1284 | ||
GenBank mRNA | AF276950.1 | - | 11 | 1169 | ||
AF276951.1 | - | 11 | 1256 | |||
AF276952.1 | - | 8 | 1012 | |||
AF454943.1 | - | 11 | 1277 | |||
AF454944.1 | - | 9 | 1149 | |||
AK029532.1 | - | 9 | 1151 | |||
AK029747.1 | - | 12 | 1398 | |||
AK049516.1 | - | 11 | 1270 | |||
AK050488.1 | - | 11 | 1223 | |||
AK157813.1 | - | 11 | 1263 | |||
AK167724.1 | - | 11 | 1415 | |||
BC019456.1 | - | 11 | 1359 | |||
BC024449.1 | - | 9 | 1132 | |||
BC038874.1 | - | 7 | 823 | |||
BC057869.1 | - | 11 | 1252 | |||
BC066821.1 | - | 9 | 911 | 30061364 |