Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323011.1 | NP_001309940.1 | 5 | 515 | ||
NM_001323012.1 | NP_001309941.1 | 5 | 522 | |||
NM_006556.3 | NP_006547.1 | 5 | 865 | |||
GenBank mRNA | AK130234.1 | - | 6 | 753 | ||
AK312803.1 | - | 5 | 824 | |||
BC006089.1 | - | 5 | 848 | 3604167 | ||
BC007694.2 | - | 5 | 778 | 3954909 | ||
BT019976.1 | - | 5 | 542 | |||
CR541998.1 | - | 5 | 542 | |||
L77213.1 | - | 5 | 577 |