Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289706.1 | NP_001276635.1 | 7 | 1137 | ||
NM_001289707.1 | NP_001276636.1 | 6 | 1062 | |||
NM_001289708.1 | NP_001276637.1 | 5 | 979 | |||
NM_020257.2 | NP_064653.1 | 6 | 1050 | |||
GenBank mRNA | AK041724.1 | - | 5 | 1765 |
Gene Symbol: Clec2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168223.1 | NP_001161695.1 | 6 | 1019 | ||
NM_001168224.1 | NP_001161696.1 | 6 | 1131 | |||
NM_027562.4 | NP_081838.1 | 7 | 1193 | |||
GenBank mRNA | AK014570.1 | - | 7 | 1195 | ||
AK137753.1 | - | 6 | 1129 | |||
BC117543.1 | - | 7 | 1183 | 40105656 |