Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027570.3 | NP_081846.3 | 9 | 1172 | ||
GenBank mRNA | AK014649.1 | - | 9 | 1178 | ||
AK029820.1 | - | 1586 | ||||
AK037996.1 | - | 482 | ||||
AY092768.1 | - | 9 | 1177 | |||
BC039155.1 | - | 9 | 1168 | 4242682 | ||
BC055443.1 | - | 9 | 1172 | 4235345 |