Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290390.1 | NP_001277319.1 | 6 | 2103 | ||
NM_001290392.1 | NP_001277321.1 | 5 | 2049 | |||
NM_001290393.1 | NP_001277322.1 | 5 | 2049 | |||
NM_027671.5 | NP_081947.2 | 6 | 2103 | |||
GenBank mRNA | AF367759.1 | - | 5 | 755 | ||
AK038698.1 | - | 5 | 2050 | |||
AK038832.1 | - | 5 | 1682 | |||
AK045676.1 | - | 5 | 1454 | |||
AK080034.1 | - | 6 | 1465 | |||
AK083241.1 | - | 6 | 1091 |