Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027769.2 | NP_082045.1 | 2, 3 | 252 | ||
GenBank mRNA | AK017357.1 | - | 2, 3 | 231 | ||
AK017651.1 | - | 2, 3 | 257 | |||
AK036664.1 | - | 2, 3 | 232 | |||
AK146324.1 | - | 2, 3 | 229 | |||
AK220348.1 | - | 2, 3 | 223 | |||
BB611055.1 | - | 2, 3 | 250 | |||
BC036983.1 | - | 2, 3 | 179 | |||
BC049226.1 | - | 13 | ||||
BC090632.1 | - | 1, 2 | 153 | 30620886 | ||
BY717432.1 | - | 2, 3 | 252 |