Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141950.1 | NP_001135422.1 | 5 | 408 | ||
NM_027797.3 | NP_082073.1 | 5 | 477 | |||
GenBank mRNA | AK017460.1 | - | 5 | 478 | ||
AK134821.1 | - | 5 | 409 | |||
AK169083.1 | - | 5 | 367 | |||
BC022756.1 | - | 5 | 352 | 4922223 |