Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027840.3 | NP_082116.1 | 2 | 234 | ||
GenBank mRNA | AF395844.1 | - | 1 | 73 | ||
AK018632.1 | - | 2 | 236 | |||
AK042068.1 | - | 2 | 209 | |||
AK157032.1 | - | 2 | 207 | |||
AK157884.1 | - | 2 | 208 | |||
AK170109.1 | - | 2 | 4312 | |||
BC039809.1 | - | 2 | 140 | 2938247 |