Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gypc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048207.1 | NP_001041672.1 | 3 | 647 | ||
GenBank mRNA | AK002769.1 | - | 3 | 477 | ||
AK082930.1 | - | 3 | 561 | |||
AK152607.1 | - | 3 | 577 | |||
AK159730.1 | - | 3 | 581 | |||
AK162472.1 | - | 2 | 380 | |||
AY234183.1 | - | 3 | 647 | |||
BC027408.1 | - | 3 | 587 | 4952917 |