Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027891.4 | NP_082167.2 | 8 | 1094 | ||
GenBank mRNA | AK004717.1 | - | 6 | 2056 | ||
AK051910.1 | - | 8 | 1042 | |||
AK084948.1 | - | 8 | 1094 | |||
AK153976.1 | - | 8 | 1065 | |||
AK154352.1 | - | 8 | 1036 | |||
AK167020.1 | - | 8 | 1043 | |||
AK172708.1 | - | 8 | 1062 | |||
BC048397.1 | - | 8 | 1073 | 6396815 |