Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168292.1 | NP_001161764.1 | 3 | 357 | ||
NM_001168293.1 | NP_001161765.1 | 2 | 672 | |||
NM_027900.4 | NP_082176.4 | 2 | 672 | |||
GenBank mRNA | AK043545.1 | - | 2 | 673 | ||
AK122416.1 | - | 2 | 215 | |||
BC117927.2 | - | 2 | 415 | 40097626 |