Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027901.2 | NP_082177.2 | 2 | 427 | ||
GenBank mRNA | AK004895.1 | - | 2 | 205 | ||
AK035792.1 | - | 2 | 193 | |||
AK043944.1 | - | 2 | 194 | |||
AK083585.1 | - | 2 | 428 | |||
AK083636.1 | - | 1 | 182 | |||
AK088596.1 | - | 2 | 195 | |||
AK129033.1 | - | 1 | 177 | |||
AK138760.1 | - | 196 | ||||
AK149771.1 | - | 2 | 195 | |||
AK151300.1 | - | 2 | 192 | |||
AK152439.1 | - | 2 | 192 | |||
AK154056.1 | - | 2 | 208 | |||
AK159938.1 | - | 2 | 194 | |||
BC034369.1 | - | 2 | 202 | 3994677 | ||
BC043100.1 | - | 2 | 415 |