Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027649.3 | NP_081925.2 | 3 | 423 | ||
GenBank mRNA | AK005865.1 | - | 3 | 424 | ||
AK016586.1 | - | 3 | 350 | |||
AK078278.1 | - | 345 | ||||
AK165435.1 | - | 2 | 211 | |||
AY336501.1 | - | 3 | 311 | |||
BC131909.1 | - | 3 | 383 | 40130186 |