Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfap3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177881.1 | NP_001171352.1 | 2, 3 | 774 | ||
NM_001177882.1 | NP_001171353.1 | 2, 3 | 542 | |||
NM_027756.5 | NP_082032.1 | 2, 3 | 666 | |||
GenBank mRNA | AK016959.1 | - | 2, 3 | 543 | ||
AK122332.1 | - | 1, 2 | 123 | |||
AK133407.1 | - | 2, 3 | 775 | |||
BC095950.1 | - | 2, 3 | 612 | 5025281 |