Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000181.3 | NP_000172.2 | 7 | 1309 | ||
NM_001284290.1 | NP_001271219.1 | 5 | 871 | |||
NM_001293104.1 | NP_001280033.1 | 6 | 1124 | |||
NM_001293105.1 | NP_001280034.1 | 5 | 981 | |||
NR_120531.1 | 7 | 1309 | ||||
XM_005250297.3 | XP_005250354.1 | 6 | 1151 | |||
XM_011516113.1 | XP_011514415.1 | 6 | 1053 | |||
XM_011516114.1 | XP_011514416.1 | 6 | 981 | |||
XM_017012091.1 | XP_016867580.1 | 5 | 900 | |||
XM_017012092.1 | XP_016867581.1 | 5 | 887 | |||
XM_017012093.1 | XP_016867582.1 | 5 | 828 | |||
GenBank mRNA | AK096764.1 | - | 1 | 1568 | ||
AK223406.1 | - | 7 | 1208 | |||
AK300223.1 | - | 7 | 1098 | |||
AK303819.1 | - | 5 | 768 | |||
AK312752.1 | - | 7 | 1225 | |||
BC014142.2 | - | 7 | 1179 | 4662011 | ||
CD014092.1 | - | 6 | 1006 | |||
CD014093.1 | - | 5 | 864 | |||
CD014094.1 | - | 7 | 1191 | |||
M15182.1 | - | 7 | 1204 |