Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164477.1 | NP_001157949.1 | 2 | 799 | ||
NM_027835.3 | NP_082111.2 | 2 | 799 | |||
GenBank mRNA | AF374384.1 | - | 2 | 708 | ||
AK037057.1 | - | 2 | 697 | |||
AK040519.1 | - | 2 | 756 | |||
AK149802.1 | - | 2 | 706 | |||
AK153018.1 | - | 2 | 706 | |||
AK156450.1 | - | 2 | 707 | |||
AK168646.1 | - | 2 | 800 | |||
AY075132.1 | - | 2 | 474 | |||
BC080200.1 | - | 2 | 704 | 3597815 | ||
EF113547.1 | - | 2 | 488 | |||
EF113548.1 | - | 2 | 488 | |||
EU084750.1 | - | 2 | 488 |