Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027890.5 | NP_082166.3 | 2, 3 | 351 | ||
GenBank mRNA | AK004703.2 | - | 2, 3 | 321 | ||
AK028817.1 | - | 2, 3 | 327 | |||
AK037059.1 | - | 2, 3 | 333 | |||
AK037127.1 | - | 2, 3 | 302 | |||
AK038944.1 | - | 2, 3 | 352 | |||
AK042469.1 | - | 2, 3 | 335 | |||
AK128959.1 | - | 2, 3 | 323 | |||
AK167155.1 | - | 2, 3 | 352 | |||
BB632717.1 | - | 2, 3 | 352 | |||
BC026837.1 | - | 2, 3 | 304 | 4222578 |