Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038616.2 | NP_001033705.2 | 5 | 1015 | ||
NM_027591.3 | NP_081867.1 | 7 | 1196 | |||
NM_029378.2 | NP_083654.1 | 5 | 1010 | |||
GenBank mRNA | AK014934.1 | - | 7 | 1197 | ||
DQ316772.1 | - | 7 | 1152 | |||
DQ316773.1 | - | 5 | 1010 | |||
DQ316774.1 | - | 5 | 927 |