Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam71d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027597.4 | NP_081873.2 | 5 | 805 | ||
NM_029069.1 | NP_083345.1 | 5 | 810 | |||
GenBank mRNA | AB473726.1 | - | 3 | 485 | ||
AK014851.1 | - | 5 | 811 | |||
AK015802.1 | - | 5 | 657 | |||
BC051149.1 | - | 5 | 665 | 6707726 |