Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027629.3 | NP_081905.1 | 14 | 4079 | ||
GenBank mRNA | AK033119.1 | - | 1 | 1294 | ||
AK039189.1 | - | 14 | 4043 | |||
AK045551.1 | - | 1 | 1286 |