Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMBS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000190.3 | NP_000181.2 | 10 | 792 | ||
NM_001024382.1 | NP_001019553.1 | 10 | 694 | |||
NM_001258208.1 | NP_001245137.1 | 10 | 792 | |||
NM_001258209.1 | NP_001245138.1 | 10 | 883 | |||
XM_005271531.1 | XP_005271588.1 | 10 | 870 | |||
XM_005271532.1 | XP_005271589.1 | 10 | 848 | |||
XM_005271533.3 | XP_005271590.1 | 9 | 744 | |||
XM_011542796.1 | XP_011541098.1 | 9 | 1124 | |||
XM_017017629.1 | XP_016873118.1 | 10 | 1258 | |||
GenBank mRNA | AK000628.1 | - | 10 | 875 | ||
AK131072.1 | - | 10 | 755 | |||
AK290275.1 | - | 10 | 788 | |||
AK309544.1 | - | 10 | 929 | |||
AK310343.1 | - | 787 | ||||
BC000520.2 | - | 10 | 786 | 2822949 | ||
BC008149.1 | - | 10 | 747 | 4178944 | ||
BC019323.1 | - | 10 | 786 | 2822949 | ||
BX647328.1 | - | 9 | 923 | |||
X04217.1 | - | 10 | 665 |