Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027721.2 | NP_081997.1 | 5 | 512 | ||
GenBank mRNA | AK017114.1 | - | 5 | 513 | ||
BC056963.1 | - | 5 | 459 |