Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027727.2 | NP_082003.1 | 15 | 1994 | ||
GenBank mRNA | AK007748.1 | - | 3 | 563 | ||
AK049909.1 | - | 15 | 1959 | |||
AK154794.1 | - | 15 | 1983 | |||
AK180386.1 | - | 3 | 286 | |||
AK198427.1 | - | 3 | 308 | |||
BC018220.1 | - | 11 | 1354 | 3985017 | ||
BC038276.1 | - | 15 | 1996 | 4458792 |