Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162913.1 | NP_001156385.1 | 9 | 1378 | ||
NM_027890.5 | NP_082166.3 | 11 | 1738 | |||
GenBank mRNA | AK004703.2 | - | 11 | 1708 | ||
AK028817.1 | - | 11 | 1714 | |||
AK033823.1 | - | 9 | 1351 | |||
AK037059.1 | - | 11 | 1720 | |||
AK037127.1 | - | 11 | 1689 | |||
AK038944.1 | - | 9 | 2006 | |||
AK042469.1 | - | 11 | 1722 | |||
AK128959.1 | - | 11 | 1710 | |||
AK167155.1 | - | 10 | 1920 | |||
BC026837.1 | - | 11 | 1691 | 4222578 | ||
BC066833.1 | - | 8 | 1537 |