Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168292.1 | NP_001161764.1 | 6 | 606 | ||
NM_001168293.1 | NP_001161765.1 | 5 | 921 | |||
NM_027900.4 | NP_082176.4 | 5 | 921 | |||
GenBank mRNA | AK043545.1 | - | 5 | 922 | ||
AK122416.1 | - | 5 | 464 | |||
BC043083.1 | - | 3 | 218 | |||
BC053099.1 | - | 1 | 25 | |||
BC117927.2 | - | 5 | 664 | 40097626 |