Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027905.2 | NP_082181.2 | 5 | 1266 | ||
GenBank mRNA | AK005048.1 | - | 5 | 1266 | ||
AK054432.1 | - | 5 | 1228 | |||
AK088096.1 | - | 5 | 1233 | |||
AK161059.1 | - | 5 | 1217 | |||
BC048849.1 | - | 2 | 289 | |||
BC117817.1 | - | 6 | 781 | |||
BC117818.1 | - | 6 | 814 | |||
BC141293.1 | - | 5 | 1335 | 9055905 |