Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163729.2 | NP_001157201.2 | 11 | 1567 | ||
NM_027910.3 | NP_082186.2 | 12 | 1720 | |||
GenBank mRNA | AB053465.1 | - | 12 | 1591 | ||
AJ278127.1 | - | 12 | 1587 | |||
AK004964.1 | - | 12 | 1697 | |||
AK141673.1 | - | 11 | 1552 | |||
AK170623.1 | - | 12 | 1592 | |||
AK180646.1 | - | 1 | 29 | |||
AK217217.1 | - | 1 | 29 | |||
BC018154.1 | - | 12 | 1688 | 3983436 |