Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027988.4 | NP_082264.2 | 6 | 1470 | ||
GenBank mRNA | AF399754.1 | - | 5 | 456 | ||
AF539797.1 | - | 5 | 456 | |||
AK009605.1 | - | 5 | 565 | |||
AK088226.1 | - | 6 | 1429 |