Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028019.3 | NP_082295.1 | 1, 2 | 392 | ||
GenBank mRNA | AK010429.1 | - | 1, 2 | 394 | ||
AK150921.1 | - | 1, 2 | 393 | |||
AK152096.1 | - | 1, 2 | 385 | |||
AK188660.1 | - | 68 | ||||
AK189150.1 | - | 68 | ||||
BC096385.1 | - | 1, 2 | 386 | 30744704 | ||
BC132307.1 | - | 1, 2 | 388 | 40130584 | ||
BC138303.1 | - | 1, 2 | 388 | 8861322 | ||
BC145236.1 | - | 1, 2 | 387 |