Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028044.2 | NP_082320.1 | 2 | 398 | ||
GenBank mRNA | AK005460.1 | - | 2 | 400 | ||
AK051121.1 | - | 2 | 395 | |||
AK051628.1 | - | 2 | 416 | |||
AK081672.1 | - | 2 | 314 | |||
AK159555.1 | - | 2 | 443 | |||
AK159871.1 | - | 2 | 428 | |||
AK166578.1 | - | 2 | 406 | |||
AK167201.1 | - | 2 | 419 | |||
AK167613.1 | - | 2 | 416 | |||
AK169021.1 | - | 2 | 423 | |||
AK169098.1 | - | 2 | 443 | |||
AK169410.1 | - | 2 | 373 | |||
AK179383.1 | - | 2 | 129 | |||
AK180041.1 | - | 1 | 148 | |||
AK184231.1 | - | 2 | 134 | |||
AK208457.1 | - | 1 | 98 | |||
AK212516.1 | - | 1 | 47 | |||
AK216318.1 | - | 2 | 129 | |||
BC055711.1 | - | 2 | 399 | 6415164 | ||
BC085268.1 | - | 2 | 176 | 6487559 |