Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163032.1 | NP_001156504.1 | 3 | 565 | ||
NM_028052.4 | NP_082328.3 | 4 | 459 | |||
GenBank mRNA | AK005132.1 | - | 4 | 380 | ||
AK032442.1 | - | 3 | 467 | |||
AK033999.1 | - | 3 | 539 | |||
AK045705.1 | - | 3 | 453 | |||
AK049661.1 | - | 3 | 411 | |||
AK078223.1 | - | 4 | 460 | |||
BC026512.1 | - | 4 | 406 | 4527827 | ||
CD354682.1 | - | 4 | 664 |