Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028060.3 | NP_082336.3 | 1 | 344 | ||
GenBank mRNA | AK005182.1 | - | 1 | 140 | ||
BC019202.1 | - | 1 | 145 | |||
BC055843.1 | - | 1 | 341 | 3991126 | ||
CD551889.1 | - | 1 | 344 | |||
CD554029.1 | - | 1 | 119 |