Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF361644.1 | - | 2 | 288 | ||
AK005532.1 | - | 2 | 271 | |||
AK031742.1 | - | 2 | 126 | |||
AK049358.1 | - | 2 | 274 | |||
AK085617.1 | - | 2 | 281 | |||
AK086940.1 | - | 2 | 221 | |||
AK143428.1 | - | 2 | 264 | |||
BC013549.1 | - | 2 | 290 | 4222133 |