Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cotl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028071.3 | NP_082347.1 | 3, 4 | 493 | ||
GenBank mRNA | AK007994.1 | - | 3, 4 | 455 | ||
AK008085.1 | - | 3, 4 | 455 | |||
AK010052.1 | - | 3, 4 | 455 | |||
AK039644.1 | - | 3, 4 | 455 | |||
AK148854.1 | - | 3, 4 | 459 | |||
AK150888.1 | - | 3, 4 | 455 | |||
AK151690.1 | - | 3, 4 | 455 | |||
AK157694.1 | - | 3, 4 | 455 | |||
AK172403.1 | - | 3, 4 | 455 | |||
AK210405.1 | - | 1, 2 | 60 | |||
AK213320.1 | - | 1, 2 | 60 | |||
BC010249.1 | - | 3, 4 | 448 | 3155195 | ||
BC011068.1 | - | 3, 4 | 448 | 3157387 | ||
CT010261.1 | - | 3, 4 | 314 |