Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028074.4 | NP_082350.3 | 4, 5 | 691 | ||
GenBank mRNA | AK049311.1 | - | 4, 5 | 693 | ||
AK169816.1 | - | 4, 5 | 637 | |||
AK171730.1 | - | 4, 5 | 560 | |||
AK209812.1 | - | 1 | 15 | |||
BC043036.1 | - | 3, 4 | 448 | 5698965 |