Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000227.4 | NP_000218.3 | 29 | 3827 | ||
NM_001127717.2 | NP_001121189.2 | 65 | 8702 | |||
NM_001127718.2 | NP_001121190.2 | 28 | 3659 | |||
NM_198129.2 | NP_937762.2 | 66 | 8870 | |||
XM_011525978.2 | XP_011524280.1 | 66 | 8897 | |||
XM_011525979.2 | XP_011524281.1 | 66 | 8888 | |||
XM_011525980.2 | XP_011524282.1 | 66 | 8879 | |||
XM_011525981.2 | XP_011524283.1 | 65 | 8765 | |||
XM_011525982.2 | XP_011524284.1 | 64 | 8600 | |||
XM_017025743.1 | XP_016881232.1 | 49 | 6581 | |||
XM_017025744.1 | XP_016881233.1 | 39 | 5262 | |||
GenBank mRNA | AB107369.1 | - | 66 | 8629 | ||
AB208853.1 | - | 21 | 2855 | |||
AY327114.1 | - | 28 | 3634 | |||
AY327115.1 | - | 66 | 8629 | |||
AY327116.1 | - | 65 | 8461 | |||
BC043618.1 | - | 23 | 3207 | |||
L34155.1 | - | 29 | 3769 | |||
X84900.1 | - | 31 | 4048 |