Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000228.2 | NP_000219.2 | 21, 22 | 3380 | ||
NM_001017402.1 | NP_001017402.1 | 20, 21 | 3626 | |||
NM_001127641.1 | NP_001121113.1 | 21, 22 | 3354 | |||
XM_005273124.4 | XP_005273181.1 | 21, 22 | 3562 | |||
XM_017001272.1 | XP_016856761.1 | 20, 21 | 3790 | |||
GenBank mRNA | AY035783.1 | - | 20, 21 | 3236 | ||
BC075838.1 | - | 20, 21 | 3411 | 6291096 | ||
D37766.1 | - | 21, 22 | 3317 | |||
KU178159.1 | - | 20, 21 | 3236 | |||
KU178160.1 | - | 4, 5 | 274 | |||
L25541.1 | - | 21, 22 | 3350 |