Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Endod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028013.3 | NP_082289.2 | 2 | 2443 | ||
GenBank mRNA | AK078056.1 | - | 2 | 2194 | ||
AK086102.1 | - | 2 | 2201 | |||
AK173036.1 | - | 2 | 2443 | |||
BC096490.1 | - | 2 | 2174 | 6439018 |