Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028041.2 | NP_082317.1 | 15 | 1766 | ||
GenBank mRNA | AF319547.1 | - | 15 | 2126 | ||
AK028274.1 | - | 15 | 1780 | |||
AK031534.1 | - | 15 | 1776 | |||
AK031679.1 | - | 15 | 1784 | |||
AK147715.1 | - | 15 | 1781 | |||
AK152989.1 | - | 15 | 1782 | |||
AK155138.1 | - | 15 | 1778 | |||
AK167603.1 | - | 15 | 1780 | |||
AK168455.1 | - | 15 | 1780 | |||
AK169255.1 | - | 15 | 1779 | |||
AK170967.1 | - | 15 | 1778 | |||
AK184886.1 | - | 3 | 230 | |||
BC021900.1 | - | 6 | 771 | |||
BC043699.1 | - | 15 | 1766 | 5344158 | ||
BC055026.1 | - | 11 | 1209 | |||
BC062920.1 | - | 15 | 1769 | |||
BC071251.1 | - | 9 | 1063 |