Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136237.1 | NP_001129709.1 | 4 | 583 | ||
NM_028051.3 | NP_082327.2 | 3 | 669 | |||
NM_028092.3 | NP_082368.1 | 3 | 545 | |||
GenBank mRNA | AK007473.1 | - | 3 | 670 | ||
AK008448.1 | - | 3 | 546 | |||
BC028990.1 | - | 4 | 544 | 5041383 | ||
CA492656.1 | - | 4 | 623 |