Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164827.1 | NP_001158299.1 | 4, 5 | 751 | ||
NM_028054.3 | NP_082330.2 | 4, 5 | 751 | |||
GenBank mRNA | AK005386.1 | - | 4, 5 | 515 | ||
BC099429.1 | - | 4, 5 | 444 | 30790629 |