Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000234.2 | NP_000225.1 | 28 | 3240 | ||
NM_001289063.1 | NP_001275992.1 | 27 | 3147 | |||
NM_001289064.1 | NP_001275993.1 | 26 | 3036 | |||
NM_001320970.1 | NP_001307899.1 | 28 | 3237 | |||
NM_001320971.1 | NP_001307900.1 | 27 | 3150 | |||
NR_110296.1 | 28 | 4240 | ||||
NR_135497.1 | 28 | 4243 | ||||
NR_135498.1 | 26 | 4072 | ||||
NR_135499.1 | 27 | 4153 | ||||
NR_135500.1 | 28 | 4420 | ||||
NR_135501.1 | 26 | 3061 | ||||
XM_017026809.1 | XP_016882298.1 | 26 | 2769 | |||
GenBank mRNA | AB208791.1 | - | 28 | 3963 | ||
AK225863.1 | - | 17 | 1595 | |||
AK297296.1 | - | 26 | 2747 | |||
AK300370.1 | - | 27 | 2888 | |||
AK300607.1 | - | 23 | 2576 | |||
BC108318.1 | - | 28 | 3240 | 6141535 | ||
BC110622.2 | - | 27 | 3892 | 40035687 | ||
M36067.1 | - | 28 | 2940 |